Merck
  • Blood Plasma Metabolomics Predicts Pregnancy in Holstein Cattle Transferred with Fresh and Vitrified/Warmed Embryos Produced in Vitro.
Blood Plasma Metabolomics Predicts Pregnancy in Holstein Cattle Transferred with Fresh and Vitrified/Warmed Embryos Produced in Vitro.
Journal of proteome research (2020-01-25)
Enrique Gómez, Pascal Salvetti, Julie Gatien, Susana Carrocera, David Martín-González, Marta Muñoz
ABSTRACT
Metabolomics may identify biomarkers in blood that differentiate pregnant from open embryo recipients. Fresh and vitrified/warmed, in vitro-produced embryos were transferred to Holstein recipients (discovery group). Recipient blood plasma collected on Day-0 (estrus) and Day-7 (before embryo transfer) were analyzed by nuclear magnetic resonance (N = 36 metabolites quantified). Metabolites whose concentrations differed between open and pregnant recipients were analyzed [(P < 0.05); false discovery rate (FDR) (P < 0.05)]. Biomarkers were identified in Day-7 plasma (receiver operator characteristic-area under curve (ROC-AUC) > 0.650; t-test P < 0.05; random forests, mean decrease accuracy) and cross-validated in independent Holstein, beef, and crossbred recipients (overall classification accuracy -OCA-; P < 0.05). Recipients with fresh embryos showed N = 6 biomarkers consistently on Day-40, Day-62, and at birth. Recipients with vitrified embryos showed N = 5 biomarkers on Day-40 and Day-62 but only one biomarker at birth. The most predictive biomarkers identified at birth within fresh embryos were oxoglutaric acid (ROC-AUC = 0.709; OCA = 0.812) and ornithine (ROC-AUC = 0.731; OCA = 0.727), while l-glycine was identified in vitrified embryos (ROC-AUC = 0.796; OCA = 0.667) together with other predictive biomarkers not identified at birth (Day-62: l-glutamine ROC-AUC = 0.757; OCA = 0.767) and l-lysine (Day-62: ROC-AUC = 0.680; OCA = 0.767). Pathway enrichment analysis distinguished between pregnant recipients for fresh (enriched energy oxidative metabolism from fat) and vitrified (lower lipid metabolism) embryos. Metabolomics can select individuals that will become pregnant in a defined cycle.