Merck
  • The impact of phenanthrene on membrane phospholipids and its biodegradation by Sphingopyxis soli.
The impact of phenanthrene on membrane phospholipids and its biodegradation by Sphingopyxis soli.
Ecotoxicology and environmental safety (2020-02-03)
Jong Cheol Shon, Young Ji Noh, Young Sang Kwon, Jong-Hwan Kim, Zhexue Wu, Jong-Su Seo
ABSTRACT
The direct interactions of bacterial membranes and polycyclic aromatic hydrocarbons (PAHs) strongly influence the biological processes, such as metabolic activity and uptake of substrates due to changes in membrane lipids. However, the elucidation of adaptation mechanisms as well as membrane phospholipid alterations in the presence of phenanthrene (PHE) from α-proteobacteria has not been fully explored. This study was conducted to define the degradation efficiency of PHE by Sphingopyxis soli strain KIT-001 in a newly isolated from Jeonju river sediments and to characterize lipid profiles in the presence of PHE in comparison to cells grown on glucose using quantitative lipidomic analysis. This strain was able to respectively utilize 1-hydroxy-2-naphthoic acid and salicylic acid as sole carbon source and approximately 90% of PHE (50 mg/L) was rapidly degraded via naphthalene route within 1 day incubation. In the cells grown on PHE, strain KIT-001 appeared to dynamically change profiles of metabolite and lipid in comparison to cells grown on glucose. The levels of primary metabolites, phosphatidylethanolamines (PE), and phosphatidic acids (PA) were significantly decreased, whereas the levels of phosphatidylcholines (PC) and phosphatidylglycerols (PG) were significantly increased. The adaptation mechanism of Sphingopyxis sp. regarded mainly the accumulation of bilayer forming lipids and anionic lipids to adapt more quickly under restricted nutrition and toxicity condition. Hence, these findings are conceivable that strain KIT-001 has a good adaptive ability and biodegradation for PHE through the alteration of phospholipids, and will be helpful for applications for effective bioremediation of PAHs-contaminated sites.
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
1-Hydroxy-2-naphthoic acid, ≥97.0%
Sigma-Aldrich
2-Carboxybenzaldehyde, 97%
Sigma-Aldrich
2-Hydroxy-1-naphthoic acid, 98%
Sigma-Aldrich
1-Hydroxy-2-naphthaldehyde, AldrichCPR