Merck
  • Characterization of Potent Odorants Causing a Pickle-like Off-Odor in Moutai-Aroma Type Baijiu by Comparative Aroma Extract Dilution Analysis, Quantitative Measurements, Aroma Addition, and Omission Studies.
Characterization of Potent Odorants Causing a Pickle-like Off-Odor in Moutai-Aroma Type Baijiu by Comparative Aroma Extract Dilution Analysis, Quantitative Measurements, Aroma Addition, and Omission Studies.
Journal of agricultural and food chemistry (2020-01-21)
Lulu Wang, Shanshan Fan, Yan Yan, Liang Yang, Shuang Chen, Yan Xu
ABSTRACT
A potent unpleasant aroma presenting a pickle-like off-odor in Moutai-aroma type Baijiu was studied by comparative aroma extract dilution analysis (AEDA). Two Moutai-aroma type Baijiu samples, one having no off-odor (A) and the other one presenting pickle-like off-odor (B), were selected for chemical analysis and sensory evaluation. The aroma compounds were isolated by headspace solid-phase microextraction and liquid-liquid extraction. AEDA, quantitative analysis, and odor activity value calculation were performed in both the A and B samples. The main differences between the two samples were obtained for 12 compounds, presenting significantly higher concentrations in the off-odor sample. A total of 30 Moutai-aroma type Baijiu samples having different intensities of pickle-like sensory defect were analyzed to confirm the differences. An aroma addition test indicated that the 12 compounds with higher concentrations contributed to the pickle-like off-odor when spiked into sample A. Finally, a triangle test involving omission of the aroma compounds from the spiked A sample proved that 2-methyl-3-furanthiol, methional, methyl 2-methyl-3-furyl disulfide, dimethyl trisulfide, 2-furfurylthiol, methanethiol, dimethyl disulfide, and bis(2-methyl-3-furyl)disulfide with higher concentrations were generally responsible for the pickle-like off-odor in Moutai-aroma type Baijiu.
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylpentanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Nonanoic acid, natural, 98%, FG
Sigma-Aldrich
Valeric acid, natural, ≥98%, FG
Sigma-Aldrich
Methyl 2-methyl-3-furyl disulfide, ≥98%, FG
Sigma-Aldrich
Ethyl phenylacetate, ReagentPlus®, 99%