Merck
  • Serial femtosecond crystallography on in vivo-grown crystals drives elucidation of mosquitocidal Cyt1Aa bioactivation cascade.
Serial femtosecond crystallography on in vivo-grown crystals drives elucidation of mosquitocidal Cyt1Aa bioactivation cascade.
Nature communications (2020-03-04)
Guillaume Tetreau, Anne-Sophie Banneville, Elena A Andreeva, Aaron S Brewster, Mark S Hunter, Raymond G Sierra, Jean-Marie Teulon, Iris D Young, Niamh Burke, Tilman A Grünewald, Joël Beaudouin, Irina Snigireva, Maria Teresa Fernandez-Luna, Alister Burt, Hyun-Woo Park, Luca Signor, Jayesh A Bafna, Rabia Sadir, Daphna Fenel, Elisabetta Boeri-Erba, Maria Bacia, Ninon Zala, Frédéric Laporte, Laurence Després, Martin Weik, Sébastien Boutet, Martin Rosenthal, Nicolas Coquelle, Manfred Burghammer, Duilio Cascio, Michael R Sawaya, Mathias Winterhalter, Enrico Gratton, Irina Gutsche, Brian Federici, Jean-Luc Pellequer, Nicholas K Sauter, Jacques-Philippe Colletier
ABSTRACT
Cyt1Aa is the one of four crystalline protoxins produced by mosquitocidal bacterium Bacillus thuringiensis israelensis (Bti) that has been shown to delay the evolution of insect resistance in the field. Limiting our understanding of Bti efficacy and the path to improved toxicity and spectrum has been ignorance of how Cyt1Aa crystallizes in vivo and of its mechanism of toxicity. Here, we use serial femtosecond crystallography to determine the Cyt1Aa protoxin structure from sub-micron-sized crystals produced in Bti. Structures determined under various pH/redox conditions illuminate the role played by previously uncharacterized disulfide-bridge and domain-swapped interfaces from crystal formation in Bti to dissolution in the larval mosquito midgut. Biochemical, toxicological and biophysical methods enable the deconvolution of key steps in the Cyt1Aa bioactivation cascade. We additionally show that the size, shape, production yield, pH sensitivity and toxicity of Cyt1Aa crystals grown in Bti can be controlled by single atom substitution.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 150,000
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 3,000-5,000
Pricing and availability is not currently available.