Merck
  • Proteome Instability Is a Therapeutic Vulnerability in Mismatch Repair-Deficient Cancer.
Proteome Instability Is a Therapeutic Vulnerability in Mismatch Repair-Deficient Cancer.
Cancer cell (2020-02-29)
Daniel J McGrail, Jeannine Garnett, Jun Yin, Hui Dai, David J H Shih, Truong Nguyen Anh Lam, Yang Li, Chaoyang Sun, Yongsheng Li, Rosemarie Schmandt, Ji Yuan Wu, Limei Hu, Yulong Liang, Guang Peng, Eric Jonasch, David Menter, Melinda S Yates, Scott Kopetz, Karen H Lu, Russell Broaddus, Gordon B Mills, Nidhi Sahni, Shiaw-Yih Lin
ABSTRACT
Deficient DNA mismatch repair (dMMR) induces a hypermutator phenotype that can lead to tumorigenesis; however, the functional impact of the high mutation burden resulting from this phenotype remains poorly explored. Here, we demonstrate that dMMR-induced destabilizing mutations lead to proteome instability in dMMR tumors, resulting in an abundance of misfolded protein aggregates. To compensate, dMMR cells utilize a Nedd8-mediated degradation pathway to facilitate clearance of misfolded proteins. Blockade of this Nedd8 clearance pathway with MLN4924 causes accumulation of misfolded protein aggregates, ultimately inducing immunogenic cell death in dMMR cancer cells. To leverage this immunogenic cell death, we combined MLN4924 treatment with PD1 inhibition and found the combination was synergistic, significantly improving efficacy over either treatment alone.
Sigma-Aldrich
(S)-SNAP-5114, ≥98% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting human MSH6
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-E6AP antibody, Mouse monoclonal, clone E6AP-330, purified from hybridoma cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human MSH2
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
MISSION® esiRNA, targeting human MLH1
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
MISSION® esiRNA, targeting human MSH3