Merck
  • Metal Exchange in the Interprotein ZnII -Binding Site of the Rad50 Hook Domain: Structural Insights into CdII -Induced DNA-Repair Inhibition.
Metal Exchange in the Interprotein ZnII -Binding Site of the Rad50 Hook Domain: Structural Insights into CdII -Induced DNA-Repair Inhibition.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-12-18)
Michał Padjasek, Maciej Maciejczyk, Michał Nowakowski, Olga Kerber, Maciej Pyrka, Wiktor Koźmiński, Artur Krężel
ABSTRACT
CdII is a major genotoxic agent that readily displaces ZnII in a multitude of zinc proteins, abrogates redox homeostasis, and deregulates cellular metalloproteome. To date, this displacement has been described mostly for cysteine(Cys)-rich intraprotein binding sites in certain zinc finger domains and metallothioneins. To visualize how a ZnII -to-CdII swap can affect the target protein's status and thus understand the molecular basis of CdII -induced genotoxicity an intermolecular ZnII -binding site from the crucial DNA repair protein Rad50 and its zinc hook domain were examined. By using a length-varied peptide base, ZnII -to-CdII displacement in Rad50's hook domain is demonstrated to alter it in a bimodal fashion: 1) CdII induces around a two-orders-of-magnitude stabilization effect (log  K 12 Zn II =20.8 vs. log  K 12 Cd II =22.7), which defines an extremely high affinity of a peptide towards a metal ion, and 2) the displacement disrupts the overall assembly of the domain, as shown by NMR spectroscopic and anisotropy decay data. Based on the results, a new model explaining the molecular mechanism of CdII genotoxicity that underlines CdII 's impact on Rad50's dimer stability and quaternary structure that could potentially result in abrogation of the major DNA damage response pathway is proposed.
Sigma-Aldrich
Fmoc-Asp(OtBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Thr(tBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Tyr(tBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
4-Mercaptophenylacetic acid, 97%
Sigma-Aldrich
Fmoc-His(Trt)-OH, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Fmoc-Gln(Trt)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Val-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Ile-OH, ≥98.0% (T)
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
Fmoc-Leu-OH, ≥97.0%
Sigma-Aldrich
Fmoc-Phe-OH, 98%
Sigma-Aldrich
Fmoc-Ala-OH, 95%
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)
Sigma-Aldrich
Fmoc-Met-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Glu(OtBu)-OH, ≥98.0% (HPLC)