Merck
  • Sizing the Bacillus anthracis PA63 channel with nonelectrolyte poly(ethylene glycols).
Sizing the Bacillus anthracis PA63 channel with nonelectrolyte poly(ethylene glycols).
Biophysical journal (2008-07-23)
Brian J Nablo, Kelly M Halverson, Joseph W F Robertson, Tam L Nguyen, Rekha G Panchal, Rick Gussio, Sina Bavari, Oleg V Krasilnikov, John J Kasianowicz
ABSTRACT
Nonelectrolyte polymers of poly(ethylene glycol) (PEG) were used to estimate the diameter of the ion channel formed by the Bacillus anthracis protective antigen 63 (PA(63)). Based on the ability of different molecular weight PEGs to partition into the pore and reduce channel conductance, the pore appears to be narrower than the one formed by Staphylococcus aureus alpha-hemolysin. Numerical integration of the PEG sample mass spectra and the channel conductance data were used to refine the estimate of the pore's PEG molecular mass cutoff (approximately 1400 g/mol). The results suggest that the limiting diameter of the PA(63) pore is <2 nm, which is consistent with an all-atom model of the PA(63) channel and previous experiments using large ions.
Product Description
Avanti
16:0 PEG750 PE, Avanti Research - A Croda Brand 880600P, powder
Avanti
18:1 PEG750 PE, Avanti Research - A Croda Brand 880630O
Avanti
16:0 PEG550 PE, Avanti Research - A Croda Brand 880500P, powder
Avanti
18:1 PEG550 PE, Avanti Research - A Croda Brand 880530O
Avanti
18:1 PEG3000 PE, Avanti Research - A Croda Brand 880330P, powder
Avanti
18:0 PEG550 PE, Avanti Research - A Croda Brand 880520P, powder
Avanti
18:0 PEG1000 PE, Avanti Research - A Croda Brand 880720P, powder
Avanti
14:0 PEG550 PE, Avanti Research - A Croda Brand 880510P, powder
Avanti
18:1 PEG350 PE, Avanti Research - A Croda Brand 880430O
Avanti
18:0 PEG1000 PE, Avanti Research - A Croda Brand 880720C