Merck
  • Lipidomic Profiling for Serum Biomarkers in Mice Exposed to Ionizing Radiation.
Lipidomic Profiling for Serum Biomarkers in Mice Exposed to Ionizing Radiation.
Dose-response : a publication of International Hormesis Society (2020-05-05)
Jinfeng Huang, Qi Wang, Zhenhua Qi, Shixiang Zhou, Meijuan Zhou, Zhidong Wang
ABSTRACT
Radiation biodosimeters are required urgently for fast and accurate evaluation of absorbed dose for irradiated individuals. Lipidomics has appeared as a credible technique for identification and quantification of lipid for researching biomarker of diseases. We performed a lipidomic profile on mice serum at time points of 6, 24, and 72 hours after 0, 2, 5.5, 7, and 8 Gy irradiation to select radiation-responsive lipids and conducted Kyoto Encyclopedia of Genes and Genome pathway enrichment analysis to recognize the pathways and network changes. Then, Pearson correlation analysis was performed to evaluate the feasibility of radiation-responsive lipids to estimate radiation dose. Seven radiation-responsive lipids including PC (18:2/18:2), PC (18:0/18:2), Lyso PC 18:1, PC (18:0/20:4), SM (D18:0/24:1), PC (16:0/18:1), and Lyso PC 18:2 were identified in which glycerophospholipid metabolism presented as the most significant pathway, and they all presented good linear correlation with the irradiated dose. This study identified 7 radiation-responsive lipids in mice serum and certificate their feasibility of dose estimation as biodosimeters.
Product Description
Avanti
19:0 PC, Avanti Research - A Croda Brand
Avanti
19:0 PC, 1,2-dinonadecanoyl-sn-glycero-3-phosphocholine, chloroform
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Avanti
19:0 Lyso PC, Avanti Research - A Croda Brand 855776P, powder
Avanti
12:0 SM (d18:1/12:0), Avanti Research - A Croda Brand 860583P, powder
Avanti
17:0 PE, Avanti Research - A Croda Brand
Avanti
19:0 Lyso PC, Avanti Research - A Croda Brand 855776C