Merck
  • A conjunctive lipidomic approach reveals plasma ethanolamine plasmalogens and fatty acids as early diagnostic biomarkers for colorectal cancer patients.
A conjunctive lipidomic approach reveals plasma ethanolamine plasmalogens and fatty acids as early diagnostic biomarkers for colorectal cancer patients.
Expert review of proteomics (2020-04-21)
Tong Liu, Zhirong Tan, Jing Yu, Feng Peng, Jiwei Guo, Wenhui Meng, Yao Chen, Tai Rao, Zhaoqian Liu, Jingbo Peng
ABSTRACT
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in plasma lipid profiles of CRC patients (n = 101) and healthy volunteers (n = 52). Based on UHPLC-Q-TOF MS and UHPLC-QQQ MS platforms, a total of 236 lipids were structurally detected. Multivariate data analysis was conducted for biomarkers discovery.Results: A total of 11 lipid species, including 1 Glycerophosphoethanolamine (PE), 3 ethanolamine plasmalogens (PlsEtn), 1 plasmanyl glycerophosphatidylethanolamine (PE-O), 3 fatty acids (FFA), 1 Fatty acid ester of hydroxyl fatty acid (FAHFA) and 2 Diacylglycerophosphates (PA) were identified to distinguish the CRC patients at early stage from healthy controls. In addition, these potential lipid biomarkers achieved an estimated AUC=0.981 in a validation set for univariate ROC analysis.Conclusion: By combining Q-TOF MS and QQQ MS analysis, the 11 lipids exhibited good performance in differentiating early-stage CRC and healthy control. This study also demonstrated that lipidomics is a powerful tool in discovering new potential biomarkers for cancer diagnosis.
Product Description
Avanti
15:0-18:1-d7-PI, Avanti Research - A Croda Brand
Avanti
18:1-d7 Lyso PC, Avanti Research - A Croda Brand
Avanti
18:1 SM (d18:1/18:1)-d9, Avanti Research - A Croda Brand 860740L-1MG860746L-1MG
Avanti
18:1 cholesteryl-d7 ester, Avanti Research - A Croda Brand
Avanti
18:1-d7 MG, Avanti Research - A Croda Brand 791646C
Avanti
15:0-18:1-d7-PA, Avanti Research - A Croda Brand
Avanti
15:0-18:1-d7-PS, Avanti Research - A Croda Brand
Avanti
15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C