Merck
  • RUVBL1/RUVBL2 ATPase Activity Drives PAQosome Maturation, DNA Replication and Radioresistance in Lung Cancer.
RUVBL1/RUVBL2 ATPase Activity Drives PAQosome Maturation, DNA Replication and Radioresistance in Lung Cancer.
Cell chemical biology (2019-12-31)
Paul Yenerall, Amit K Das, Shan Wang, Rahul K Kollipara, Long Shan Li, Pamela Villalobos, Josiah Flaming, Yu-Fen Lin, Kenneth Huffman, Brenda C Timmons, Collin Gilbreath, Rajni Sonavane, Lisa N Kinch, Jaime Rodriguez-Canales, Cesar Moran, Carmen Behrens, Makoto Hirasawa, Takehiko Takata, Ryo Murakami, Koichi Iwanaga, Benjamin P C Chen, Nick V Grishin, Ganesh V Raj, Ignacio I Wistuba, John D Minna, Ralf Kittler
ABSTRACT
RUVBL1 and RUVBL2 (collectively RUVBL1/2) are essential AAA+ ATPases that function as co-chaperones and have been implicated in cancer. Here we investigated the molecular and phenotypic role of RUVBL1/2 ATPase activity in non-small cell lung cancer (NSCLC). We find that RUVBL1/2 are overexpressed in NSCLC patient tumors, with high expression associated with poor survival. Utilizing a specific inhibitor of RUVBL1/2 ATPase activity, we show that RUVBL1/2 ATPase activity is necessary for the maturation or dissociation of the PAQosome, a large RUVBL1/2-dependent multiprotein complex. We also show that RUVBL1/2 have roles in DNA replication, as inhibition of its ATPase activity can cause S-phase arrest, which culminates in cancer cell death via replication catastrophe. While in vivo pharmacological inhibition of RUVBL1/2 results in modest antitumor activity, it synergizes with radiation in NSCLC, but not normal cells, an attractive property for future preclinical development.
Product Description
Millipore
Klebsiella Selective Supplement, suitable for microbiology
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Corning® cell strainer, pore size 70 μm, white, sterile, pkg of (individually wrapped), pack of 50 ea, Corning 431751
Sigma-Aldrich
Anti-Pontin antibody, Mouse monoclonal, clone 5G3-11, purified from hybridoma cell culture
Sigma-Aldrich
V5 Peptide, ≥97% (HPLC), lyophilized powder
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, biotin conjugate, clone JBW301, Upstate®, from mouse