Merck
  • An Integrated Gene Expression Landscape Profiling Approach to Identify Lung Tumor Endothelial Cell Heterogeneity and Angiogenic Candidates.
An Integrated Gene Expression Landscape Profiling Approach to Identify Lung Tumor Endothelial Cell Heterogeneity and Angiogenic Candidates.
Cancer cell (2020-01-15)
Jermaine Goveia, Katerina Rohlenova, Federico Taverna, Lucas Treps, Lena-Christin Conradi, Andreas Pircher, Vincent Geldhof, Laura P M H de Rooij, Joanna Kalucka, Liliana Sokol, Melissa García-Caballero, Yingfeng Zheng, Junbin Qian, Laure-Anne Teuwen, Shawez Khan, Bram Boeckx, Els Wauters, Herbert Decaluwé, Paul De Leyn, Johan Vansteenkiste, Birgit Weynand, Xavier Sagaert, Erik Verbeken, Albert Wolthuis, Baki Topal, Wouter Everaerts, Hanibal Bohnenberger, Alexander Emmert, Dena Panovska, Frederik De Smet, Frank J T Staal, Rene J Mclaughlin, Francis Impens, Vincenzo Lagani, Stefan Vinckier, Massimiliano Mazzone, Luc Schoonjans, Mieke Dewerchin, Guy Eelen, Tobias K Karakach, Huanming Yang, Jian Wang, Lars Bolund, Lin Lin, Bernard Thienpont, Xuri Li, Diether Lambrechts, Yonglun Luo, Peter Carmeliet
ABSTRACT
Heterogeneity of lung tumor endothelial cell (TEC) phenotypes across patients, species (human/mouse), and models (in vivo/in vitro) remains poorly inventoried at the single-cell level. We single-cell RNA (scRNA)-sequenced 56,771 endothelial cells from human/mouse (peri)-tumoral lung and cultured human lung TECs, and detected 17 known and 16 previously unrecognized phenotypes, including TECs putatively regulating immune surveillance. We resolved the canonical tip TECs into a known migratory tip and a putative basement-membrane remodeling breach phenotype. Tip TEC signatures correlated with patient survival, and tip/breach TECs were most sensitive to vascular endothelial growth factor blockade. Only tip TECs were congruent across species/models and shared conserved markers. Integrated analysis of the scRNA-sequenced data with orthogonal multi-omics and meta-analysis data across different human tumors, validated by functional analysis, identified collagen modification as a candidate angiogenic pathway.
Sigma-Aldrich
Minoxidil, ≥99% (TLC)
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-SELP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
3-amino-propionitrile, AldrichCPR
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
3-Aminopropionitrile fumarate salt, metabolite
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Collagen Type I, rat tail