Merck
  • A Ca2+-dependent mechanism of neuronal survival mediated by the microtubule-associated protein p600.
A Ca2+-dependent mechanism of neuronal survival mediated by the microtubule-associated protein p600.
The Journal of biological chemistry (2013-07-19)
Camille Belzil, Gernot Neumayer, Alex P Vassilev, Kyoko L Yap, Hiroaki Konishi, Serge Rivest, Kamon Sanada, Mitsuhiko Ikura, Yoshihiro Nakatani, Minh Dang Nguyen
ABSTRACT
In acute and chronic neurodegeneration, Ca(2+) mishandling and disruption of the cytoskeleton compromise neuronal integrity, yet abnormalities in the signaling roles of cytoskeletal proteins remain largely unexplored. We now report that the microtubule-associated protein p600 (also known as UBR4) promotes neuronal survival. Following depletion of p600, glutamate-induced Ca(2+) influx through NMDA receptors, but not AMPA receptors, initiates a degenerative process characterized by endoplasmic reticulum fragmentation and endoplasmic reticulum Ca(2+) release via inositol 1,4,5-trisphosphate receptors. Downstream of NMDA receptors, p600 associates with the calmodulin·calmodulin-dependent protein kinase IIα complex. A direct and atypical p600/calmodulin interaction is required for neuronal survival. Thus, p600 counteracts specific Ca(2+)-induced death pathways through regulation of Ca(2+) homeostasis and signaling.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SP15, ascites fluid, clone SP15, Chemicon®
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®