Merck
  • Post-translational myristoylation: Fat matters in cellular life and death.
Post-translational myristoylation: Fat matters in cellular life and death.
Biochimie (2010-11-09)
Dale D O Martin, Erwan Beauchamp, Luc G Berthiaume
ABSTRACT
Myristoylation corresponds to the irreversible covalent linkage of the 14-carbon saturated fatty acid, myristic acid, to the N-terminal glycine of many eukaryotic and viral proteins. It is catalyzed by N-myristoyltransferase. Typically, the myristate moiety participates in protein subcellular localization by facilitating protein-membrane interactions as well as protein-protein interactions. Myristoylated proteins are crucial components of a wide variety of functions, which include many signalling pathways, oncogenesis or viral replication. Initially, myristoylation was described as a co-translational reaction that occurs after the removal of the initiator methionine residue. However, it is now well established that myristoylation can also occur post-translationally in apoptotic cells. Indeed, during apoptosis hundreds of proteins are cleaved by caspases and in many cases this cleavage exposes an N-terminal glycine within a cryptic myristoylation consensus sequence, which can be myristoylated. The principal objective of this review is to provide an overview on the implication of myristoylation in health and disease with a special emphasis on post-translational myristoylation. In addition, new advancements in the detection and identification of myristoylated proteins are also briefly reviewed.
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Myristic acid, analytical standard
USP
Myristic acid, United States Pharmacopeia (USP) Reference Standard
Avanti
14:0 Coenzyme A, Avanti Research - A Croda Brand 870714P, powder