Merck
  • UTX-mediated enhancer and chromatin remodeling suppresses myeloid leukemogenesis through noncatalytic inverse regulation of ETS and GATA programs.
UTX-mediated enhancer and chromatin remodeling suppresses myeloid leukemogenesis through noncatalytic inverse regulation of ETS and GATA programs.
Nature genetics (2018-05-08)
Malgorzata Gozdecka, Eshwar Meduri, Milena Mazan, Konstantinos Tzelepis, Monika Dudek, Andrew J Knights, Mercedes Pardo, Lu Yu, Jyoti S Choudhary, Emmanouil Metzakopian, Vivek Iyer, Haiyang Yun, Naomi Park, Ignacio Varela, Ruben Bautista, Grace Collord, Oliver Dovey, Dimitrios A Garyfallos, Etienne De Braekeleer, Saki Kondo, Jonathan Cooper, Berthold Göttgens, Lars Bullinger, Paul A Northcott, David Adams, George S Vassiliou, Brian J P Huntly
ABSTRACT
The histone H3 Lys27-specific demethylase UTX (or KDM6A) is targeted by loss-of-function mutations in multiple cancers. Here, we demonstrate that UTX suppresses myeloid leukemogenesis through noncatalytic functions, a property shared with its catalytically inactive Y-chromosome paralog, UTY (or KDM6C). In keeping with this, we demonstrate concomitant loss/mutation of KDM6A (UTX) and UTY in multiple human cancers. Mechanistically, global genomic profiling showed only minor changes in H3K27me3 but significant and bidirectional alterations in H3K27ac and chromatin accessibility; a predominant loss of H3K4me1 modifications; alterations in ETS and GATA-factor binding; and altered gene expression after Utx loss. By integrating proteomic and genomic analyses, we link these changes to UTX regulation of ATP-dependent chromatin remodeling, coordination of the COMPASS complex and enhanced pioneering activity of ETS factors during evolution to AML. Collectively, our findings identify a dual role for UTX in suppressing acute myeloid leukemia via repression of oncogenic ETS and upregulation of tumor-suppressive GATA programs.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture