Merck
  • SOD1 Phosphorylation by mTORC1 Couples Nutrient Sensing and Redox Regulation.
SOD1 Phosphorylation by mTORC1 Couples Nutrient Sensing and Redox Regulation.
Molecular cell (2018-05-05)
Chi Kwan Tsang, Miao Chen, Xin Cheng, Yanmei Qi, Yin Chen, Ishani Das, Xiaoxing Li, Brinda Vallat, Li-Wu Fu, Chao-Nan Qian, Hui-Yun Wang, Eileen White, Stephen K Burley, X F Steven Zheng
ABSTRACT
Nutrients are not only organic compounds fueling bioenergetics and biosynthesis, but also key chemical signals controlling growth and metabolism. Nutrients enormously impact the production of reactive oxygen species (ROS), which play essential roles in normal physiology and diseases. How nutrient signaling is integrated with redox regulation is an interesting, but not fully understood, question. Herein, we report that superoxide dismutase 1 (SOD1) is a conserved component of the mechanistic target of rapamycin complex 1 (mTORC1) nutrient signaling. mTORC1 regulates SOD1 activity through reversible phosphorylation at S39 in yeast and T40 in humans in response to nutrients, which moderates ROS level and prevents oxidative DNA damage. We further show that SOD1 activation enhances cancer cell survival and tumor formation in the ischemic tumor microenvironment and protects against the chemotherapeutic agent cisplatin. Collectively, these findings identify a conserved mechanism by which eukaryotes dynamically regulate redox homeostasis in response to changing nutrient conditions.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit
Sigma-Aldrich
4-Hydroxy-TEMPO, 95%
Sigma-Aldrich
Anti-8-Oxoguanine Antibody, clone 483.15, ascites fluid, clone 483.15, Chemicon®