Merck
  • MIGA2 Links Mitochondria, the ER, and Lipid Droplets and Promotes De Novo Lipogenesis in Adipocytes.
MIGA2 Links Mitochondria, the ER, and Lipid Droplets and Promotes De Novo Lipogenesis in Adipocytes.
Molecular cell (2019-10-20)
Christophe A C Freyre, Pascal C Rauher, Christer S Ejsing, Robin W Klemm
ABSTRACT
Physical contact between organelles is vital to the function of eukaryotic cells. Lipid droplets (LDs) are dynamic organelles specialized in lipid storage that interact physically with mitochondria in several cell types. The mechanisms coupling these organelles are, however, poorly understood, and the cell-biological function of their interaction remains largely unknown. Here, we discover in adipocytes that the outer mitochondrial membrane protein MIGA2 links mitochondria to LDs. We identify an amphipathic LD-targeting motif and reveal that MIGA2 binds to the membrane proteins VAP-A or VAP-B in the endoplasmic reticulum (ER). We find that in adipocytes MIGA2 is involved in promoting triglyceride (TAG) synthesis from non-lipid precursors. Our data indicate that MIGA2 links reactions of de novo lipogenesis in mitochondria to TAG production in the ER, thereby facilitating efficient lipid storage in LDs. Based on its presence in many tissues, MIGA2 is likely critical for lipid and energy homeostasis in a wide spectrum of cell types.
Sigma-Aldrich
Cholesterol-25,26,26,26,27,27,27-d7, 99 atom % D, 98% (CP)
Sigma-Aldrich
Hexadimethrine bromide, ≥95%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Avanti
15:0-18:1-d7-PI, Avanti Research - A Croda Brand
Avanti
C12 Glucosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860543P, powder
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
ANTI-PLIN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Avanti
15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C
Avanti
15:0-18:1-d7-PS, Avanti Research - A Croda Brand