Merck
  • Evidence of nuclear transport mechanisms in the protozoan parasite Giardia lamblia.
Evidence of nuclear transport mechanisms in the protozoan parasite Giardia lamblia.
Biochimica et biophysica acta. Molecular cell research (2019-11-02)
Gonzalo Federico Mayol, María Victoria Revuelta, Agostina Salusso, María Carolina Touz, Andrea Silvana Rópolo
ABSTRACT
Nuclear-cytoplasmic trafficking of proteins is a highly regulated process that modulates multiple biological processes in eukaryotic cells. In Giardia lamblia, shuttling has been described from the cytoplasm to nuclei of proteins during the biological cell cycle of the parasite. This suggests that a mechanism of nucleocytoplasmic transport is present and functional in G. lamblia. By means of computational biology analyses, we found that there are only two genes for nuclear transport in this parasite, named Importin α and Importin β. When these transporters were overexpressed, both localized close to the nuclear envelope, and no change was observed in trophozoite growth rate. However, during the encystation process, both transporters induced an increase in the number of cysts produced. Importazole and Ivermectin, two known specific inhibitors of importins, separately influenced the encysting process by inducing an arrest in the trophozoite stage that prevents the production of cysts. This effect was more noticeable when Ivermectin, an anti-parasitic drug, was used. Finally, we tested whether the enzyme arginine deiminase, which shuttles from the cytoplasm to the nuclei during encystation, was influenced by these transporters. We found that treatment with each of the inhibitors abrogates arginine deiminase nuclear translocation and favors perinuclear localization. This suggests that Importin α and Importin β are key transporters during the encystation process and are involved, at least, in the transport of arginine deiminase into the nuclei. Considering the effect produced by Ivermectin during growth and encystation, we postulate that this drug could be used to treat giardiasis.
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Importazole, ≥98% (HPLC)
Sigma-Aldrich
Anti-HA−FITC antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bile from bovine and ovine, bile acid mixture
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Supelco
m-Coumaric acid, analytical standard
Sigma-Aldrich
ANTI-V5 TAG antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution