Merck
  • Targeting nucleotide exchange to inhibit constitutively active G protein α subunits in cancer cells.
Targeting nucleotide exchange to inhibit constitutively active G protein α subunits in cancer cells.
Science signaling (2018-09-06)
Michael D Onken, Carol M Makepeace, Kevin M Kaltenbronn, Stanley M Kanai, Tyson D Todd, Shiqi Wang, Thomas J Broekelmann, Prabakar Kumar Rao, John A Cooper, Kendall J Blumer
ABSTRACT
Constitutively active G protein α subunits cause cancer, cholera, Sturge-Weber syndrome, and other disorders. Therapeutic intervention by targeted inhibition of constitutively active Gα subunits in these disorders has yet to be achieved. We found that constitutively active Gαq in uveal melanoma (UM) cells was inhibited by the cyclic depsipeptide FR900359 (FR). FR allosterically inhibited guanosine diphosphate-for-guanosine triphosphate (GDP/GTP) exchange to trap constitutively active Gαq in inactive, GDP-bound Gαβγ heterotrimers. Allosteric inhibition of other Gα subunits was achieved by the introduction of an FR-binding site. In UM cells driven by constitutively active Gαq, FR inhibited second messenger signaling, arrested cell proliferation, reinstated melanocytic differentiation, and stimulated apoptosis. In contrast, FR had no effect on BRAF-driven UM cells. FR promoted UM cell differentiation by reactivating polycomb repressive complex 2 (PRC2)-mediated gene silencing, a heretofore unrecognized effector system of constitutively active Gαq in UM. Constitutively active Gαq and PRC2 therefore provide therapeutic targets for UM. The development of FR analogs specific for other Gα subunit subtypes may provide novel therapeutic approaches for diseases driven by constitutively active Gα subunits or multiple G protein-coupled receptors (GPCRs) where targeting a single receptor is ineffective.
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, clone A3S, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
GSK503, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution