Merck
  • Role of alpha 1A adrenoceptor subtype in production of the positive inotropic effect mediated via myocardial alpha 1 adrenoceptors in the rabbit papillary muscle: influence of selective alpha 1A subtype antagonists WB 4101 and 5-methylurapidil.
Role of alpha 1A adrenoceptor subtype in production of the positive inotropic effect mediated via myocardial alpha 1 adrenoceptors in the rabbit papillary muscle: influence of selective alpha 1A subtype antagonists WB 4101 and 5-methylurapidil.
Naunyn-Schmiedeberg's archives of pharmacology (1992-05-01)
M Endoh, M Takanashi, I Norota
ABSTRACT
In order to elucidate the contribution of alpha 1A subtype to the positive inotropic effect mediated by myocardial alpha 1 adrenoceptors, the influence of the alpha 1A selective antagonists WB 4101 and 5-methylurapidil on the alpha 1-mediated positive inotropic effect (induced by phenylephrine in the presence of a beta adrenoceptor blocking agent bupranolol) was assessed in the isolated rabbit papillary muscle. WB 4101 (10(-9)-10(-7) mol/l) shifted the concentration-response curve of the alpha 1-mediated positive inotropic effect to the right in parallel, but the slope of Schild plot did not meet the competitive antagonism: WB 4101 shifted the curve by log one unit at 10(-9) mol/l, whereas it did not cause further shift at higher concentrations of 10(-8) and 10(-7) mol/l. WB 4101 did not affect the beta adrenoceptor-mediated positive inotropic effect. 5-Methylurapidil (10(-9) to 10(-7) mol/l) shifted the curve of alpha 1-mediated positive inotropic effect to the right and downwards in a concentration-dependent manner; the slope of Schild plot calculated at the level of 20% of the maximum response to phenylephrine was close to unity. 5-Methylurapidil at 3 x 10(-7) mol/l abolished the alpha 1-mediated positive inotropic effect. In addition, 5-methylurapidil inhibited the beta adrenoceptor-mediated positive inotropic effect in the same concentration range as it antagonized the alpha 1-mediated positive inotropic effect, indicating that 5-methylurapidil is not selective for myocardial alpha 1 adrenoceptors.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
5-Methylurapidil, solid