Merck
  • Role of Supramolecule ErpY-Like Lipoprotein of Leptospira in Thrombin-Catalyzed Fibrin Clot Inhibition and Binding to Complement Factors H and I, and Its Diagnostic Potential.
Role of Supramolecule ErpY-Like Lipoprotein of Leptospira in Thrombin-Catalyzed Fibrin Clot Inhibition and Binding to Complement Factors H and I, and Its Diagnostic Potential.
Infection and immunity (2019-09-25)
Karukriti Kaushik Ghosh, Aman Prakash, Anusua Dhara, Md Saddam Hussain, Prateek Shrivastav, Pankaj Kumar, Vinayagamurthy Balamurugan, Manish Kumar
ABSTRACT
Leptospirosis is one of the most widespread zoonoses caused by pathogenic Leptospira spp. In this study, we report that the LIC11966/ErpY-like lipoprotein is a surface-exposed outer membrane protein exclusively present in pathogenic species of Leptospira The recombinant ErpY (rErpY)-like protein is recognized by the immunoglobulins of confirmed leptospirosis sera of diverse hosts (human, bovine, and canine), suggesting the expression of the native leptospiral surface protein during infection. Circular dichroism of pure rErpY-like protein showed the secondary structural integrity to be uncompromised during the purification process. Analysis of the rErpY-like protein by native polyacrylamide gel electrophoresis, chemical cross-linking, dynamic light scattering, and field emission transmission electron microscopy demonstrated it undergoes supramolecular assembly. The rErpY-like protein can bind to diverse host extracellular matrices, and it presented a saturable and strong binding affinity (dissociation constant [KD ] of 70.45 ± 4.13 nM) to fibrinogen, a central host plasma component involved in blood clotting. In the presence of the rErpY-like supramolecule, thrombin-catalyzed fibrin clot formation is inhibited up to 7%, implying its role in inhibiting blood coagulation during Leptospira infection. In addition, binding of the rErpY-like supramolecule to complement factors H and I suggests the protein also contributes to Leptospira evading innate host defense during infection by inactivating alternative complement pathways. This study reveals that rErpY-like protein is functionally active in the supramolecular state and performs moonlighting activity under the given in vitro conditions.
Sigma-Aldrich
Chondroitin sulfate B sodium salt, from Porcine intestinal mucosa, ≥90%, lyophilized powder
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sigma-Aldrich
Anti-CFH antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Heparan sulfate sodium salt from bovine kidney
Sigma-Aldrich
Complement factor H from human plasma, 1 mg/mL in PBS, pH 7.2, ≥90% (SDS-PAGE)
Sigma-Aldrich
Complement factor I from human plasma, >90% (SDS-PAGE)
Sigma-Aldrich
Plasma, from human
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type I, rat tail
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture