Merck
  • 2-Methyltetrahydrofuran (2-MeTHF): a biomass-derived solvent with broad application in organic chemistry.
2-Methyltetrahydrofuran (2-MeTHF): a biomass-derived solvent with broad application in organic chemistry.
ChemSusChem (2012-08-14)
Vittorio Pace, Pilar Hoyos, Laura Castoldi, Pablo Domínguez de María, Andrés R Alcántara
ABSTRACT
2-Methyl-tetrahydrofuran (2-MeTHF) can be derived from renewable resources (e.g., furfural or levulinic acid) and is a promising alternative solvent in the search for environmentally benign synthesis strategies. Its physical and chemical properties, such as its low miscibility with water, boiling point, remarkable stability compared to other cyclic-based solvents such as THF, and others make it appealing for applications in syntheses involving organometallics, organocatalysis, and biotransformations or for processing lignocellulosic materials. Interestingly, a significant number of industries have also started to assess 2-MeTHF in several synthetic procedures, often with excellent results and prospects. Likewise, preliminary toxicology assessments suggest that the use of 2-MeTHF might even be extended to more processes in pharmaceutical chemistry. This Minireview describes the properties of 2-MeTHF, the state-of-the-art of its use in synthesis, and covers several outstanding examples of its application from both industry and academia.
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®