Merck
  • The balance between KIFC3 and EG5 tetrameric kinesins controls the onset of mitotic spindle assembly.
The balance between KIFC3 and EG5 tetrameric kinesins controls the onset of mitotic spindle assembly.
Nature cell biology (2019-09-05)
Shoji Hata, Ana Pastor Peidro, Marko Panic, Peng Liu, Enrico Atorino, Charlotta Funaya, Ursula Jäkle, Gislene Pereira, Elmar Schiebel
ABSTRACT
One of the first steps in mitotic spindle assembly is the dissolution of the centrosome linker followed by centrosome separation driven by EG5, a tetrameric plus-end-directed member of the kinesin-5 family. However, even in the absence of the centrosome linker, the two centrosomes are kept together by an ill-defined microtubule-dependent mechanism. Here we show that KIFC3, a minus-end-directed kinesin-14, provides microtubule-based centrosome cohesion. KIFC3 forms a homotetramer that pulls the two centrosomes together via a specific microtubule network. At mitotic onset, KIFC3 activity becomes the main driving force of centrosome cohesion to prevent premature spindle formation after linker dissolution as it counteracts the increasing EG5-driven pushing forces. KIFC3 is eventually inactivated by NEver in mitosis-related Kinase 2 (NEK2) to enable EG5-driven bipolar spindle assembly. We further show that persistent centrosome cohesion in mitosis leads to chromosome mis-segregation. Our findings reveal a mechanism of spindle assembly that is evolutionary conserved from yeast to humans.
Sigma-Aldrich
(+)-S-Trityl-L-cysteine, 97%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK5RAP2
Sigma-Aldrich
MISSION® esiRNA, targeting human PDE4DIP
Sigma-Aldrich
MISSION® esiRNA, targeting human KIFC1
Sigma-Aldrich
Aurora-A Inhibitor I, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human KIFC2
Sigma-Aldrich
MISSION® esiRNA, targeting human NIN
Sigma-Aldrich
MISSION® esiRNA, targeting human AKAP9 (1)
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF25
Sigma-Aldrich
MISSION® esiRNA, targeting human KIFC3
Sigma-Aldrich
MISSION® esiRNA, targeting human DYNC2H1
Sigma-Aldrich
MISSION® esiRNA, targeting human DYNC1H1