Merck
  • Synthesis of ZnO nanoparticles using insulin-rich leaf extract: Anti-diabetic, antibiofilm and anti-oxidant properties.
Synthesis of ZnO nanoparticles using insulin-rich leaf extract: Anti-diabetic, antibiofilm and anti-oxidant properties.
Journal of photochemistry and photobiology. B, Biology (2019-07-05)
Viswanathan Vinotha, Arokiadhas Iswarya, Rajagopalan Thaya, Marimuthu Govindarajan, Naiyf S Alharbi, Shine Kadaikunnan, Jamal M Khaled, Mohammed N Al-Anbr, Baskaralingam Vaseeharan
ABSTRACT
Here, we report the novel fabrication of ZnO nanoparticles using the Costus igneus leaf extract. Gas chromatography-mass spectrometry (GC-MS) and proton nuclear magnetic resonance (1H NMR) spectroscopy to determine the bioactive components present in the plant extract. The synthesis of Ci-ZnO NPs (C. igneus- coated zinc oxide nanoparticles) was accomplished using a cost-effective and simple technique. Ci-ZnO NPs were specified using UV-visible spectroscopy, FTIR, XRD, and TEM. Ci-ZnO NPs was authenticated by UV-Vis and exhibited a peak at 365 nm. The XRD spectra proved the crystalline character of the Ci-ZnO NPs synthesized as hexagonal wurtzite. The FTIR spectrum illustrated the presence of possible functional groups present in Ci-ZnO NPs. The TEM micrograph showed evidence of the presence of a hexagonal organization with a size of 26.55 nm typical of Ci-ZnO NPs. The α-amylase and α-glucosidase inhibition assays demonstrated antidiabetic activity of Ci-ZnO NPs (74 % and 82 %, respectively), and the DPPH [2,2-diphenyl-1-picrylhydrazyl hydrate] assay demonstrated the antioxidant activity of the nanoparticles (75%) at a concentration of 100 μg/ml. The Ci-ZnO NPs exhibited promising antibacterial and biofilm inhibition activity against the pathogenic bacteria Streptococcus mutans, Lysinibacillus fusiformis, Proteus vulgaris, and Vibrio parahaemolyticus. Additionally, the Ci-ZnO NPs showed biocompatibility with mammalian RBCs with minimum hemolytic activity (0.633 % ± 0.005 %) at a concentration of 200 μg/ml.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Acridine Orange base, Dye content 75 %