Merck
  • GAPDH Overexpression in the T Cell Lineage Promotes Angioimmunoblastic T Cell Lymphoma through an NF-κB-Dependent Mechanism.
GAPDH Overexpression in the T Cell Lineage Promotes Angioimmunoblastic T Cell Lymphoma through an NF-κB-Dependent Mechanism.
Cancer cell (2019-08-27)
Laura Mondragón, Rana Mhaidly, Gian Marco De Donatis, Marie Tosolini, Pascal Dao, Anthony R Martin, Caroline Pons, Johanna Chiche, Marie Jacquin, Véronique Imbert, Emma Proïcs, Laurent Boyer, Anne Doye, Frédéric Luciano, Jaap G Neels, Frédéric Coutant, Nicole Fabien, Laura Sormani, Camila Rubio-Patiño, Jozef P Bossowski, Florian Muller, Sandrine Marchetti, Elodie Villa, Jean-François Peyron, Philippe Gaulard, François Lemonnier, Vahid Asnafi, Laurent Genestier, Rachid Benhida, Jean-Jacques Fournié, Thierry Passeron, Jean-Ehrland Ricci, Els Verhoeyen
ABSTRACT
GAPDH is emerging as a key player in T cell development and function. To investigate the role of GAPDH in T cells, we generated a transgenic mouse model overexpressing GAPDH in the T cell lineage. Aged mice developed a peripheral Tfh-like lymphoma that recapitulated key molecular, pathological, and immunophenotypic features of human angioimmunoblastic T cell lymphoma (AITL). GAPDH induced non-canonical NF-κB pathway activation in mouse T cells, which was strongly activated in human AITL. We developed a NIK inhibitor to reveal that targeting the NF-κB pathway prolonged AITL-bearing mouse survival alone and in combination with anti-PD-1. These findings suggest the therapeutic potential of targeting NF-κB signaling in AITL and provide a model for future AITL therapeutic investigations.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate diethyl acetal barium salt
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ≥98.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K14
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human NFKB2
Sigma-Aldrich
D-Glyceraldehyde 3-phosphate solution, 8-13 mg/mL in H2O
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Rotenone, ≥95%