Merck
  • A modular paper-and-plastic device for tuberculosis nucleic acid amplification testing in limited-resource settings.
A modular paper-and-plastic device for tuberculosis nucleic acid amplification testing in limited-resource settings.
Scientific reports (2019-10-28)
Navjot Kaur, Joy S Michael, Bhushan J Toley
ABSTRACT
We present a prototype for conducting rapid, inexpensive and point-of-care-compatible nucleic acid amplification tests (NAATs) for tuberculosis (TB). The fluorescent isothermal paper-and-plastic NAAT (FLIPP-NAAT) uses paper-based loop mediated isothermal amplification (LAMP) for DNA detection. The cost of materials required to build a 12-test-zone device is $0.88 and the cost of reagents per reaction is $0.43. An inexpensive imaging platform enables filter-free fluorescence detection of amplified DNA using a cell-phone camera. FLIPP-NAAT can be operated by an untrained user and only requires a regular laboratory incubator as ancillary equipment. All reagents can be dry-stored in the device, facilitating storage and transportation without cold chains. The device design is modular and the assay demonstrated high specificity to Mycobacterium tuberculosis (Mtb), analytical sensitivity of the order of 10 copies of Mtb gDNA, and tolerance to complex samples. The clinical sensitivity and specificity of sputum-based FLIPP NAAT tests were 100% (zero false negatives) and 68.75% (five false positives), respectively (N = 30), using Xpert MTB/RIF assay as the reference standard. FLIPP-NAAT has the potential to provide affordable and accessible molecular diagnostics for TB in low- and middle-income countries, when used in conjunction with an appropriate sample preparation technique. Although demonstrated for the detection of TB, FLIPP-NAAT is a platform technology for amplification of any nucleic acid sequence.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
SYBR® Green I nucleic acid gel stain, 10,000 × in DMSO
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Millipore
Egg Yolk Emulsion, suitable for microbiology