Merck
  • Lipidomic Profiling Reveals Significant Perturbations of Intracellular Lipid Homeostasis in Enterovirus-Infected Cells.
Lipidomic Profiling Reveals Significant Perturbations of Intracellular Lipid Homeostasis in Enterovirus-Infected Cells.
International journal of molecular sciences (2019-11-30)
Bingpeng Yan, Zijiao Zou, Hin Chu, Gabriella Chan, Jessica Oi-Ling Tsang, Pok-Man Lai, Shuofeng Yuan, Cyril Chik-Yan Yip, Feifei Yin, Richard Yi-Tsun Kao, Kong-Hung Sze, Susanna Kar-Pui Lau, Jasper Fuk-Woo Chan, Kwok-Yung Yuen
ABSTRACT
Enterovirus A71 (EV-A71) and coxsackievirus A16 (CV-A16) are the most common causes of hand, foot, and mouth disease. Severe EV-A71 and CV-A16 infections may be associated with life-threatening complications. However, the pathogenic mechanisms underlying these severe clinical and pathological features remain incompletely understood. Lipids are known to play critical roles in multiple stages of the virus replication cycle. The specific lipid profile induced upon virus infection is required for optimal virus replication. The perturbations in the host cell lipidomic profiles upon enterovirus infection have not been fully characterized. To this end, we performed ultra-high performance liquid chromatography-electrospray ionization-quadrupole-time of flight-mass spectrometry (UPLC-ESI-Q-TOF-MS)-based lipidomics to characterize the change in host lipidome upon EV-A71 and CV-A16 infections. Our results revealed that 47 lipids within 11 lipid classes were significantly perturbed after EV-A71 and CV-A16 infection. Four polyunsaturated fatty acids (PUFAs), namely, arachidonic acid (AA), docosahexaenoic acid (DHA), docosapentaenoic acid (DPA), and eicosapentaenoic acid (EPA), were consistently upregulated upon EV-A71 and CV-A16 infection. Importantly, exogenously supplying three of these four PUFAs, including AA, DHA, and EPA, in cell cultures significantly reduced EV-A71 and CV-A16 replication. Taken together, our results suggested that enteroviruses might specifically modulate the host lipid pathways for optimal virus replication. Excessive exogenous addition of lipids that disrupted this delicate homeostatic state could prevent efficient viral replication. Precise manipulation of the host lipid profile might be a potential host-targeting antiviral strategy for enterovirus infection.
Product Description
Avanti
C17 Ceramide (d18:1/17:0), Avanti Research - A Croda Brand
Avanti
cholesterol-d7, Avanti Research - A Croda Brand
Avanti
Sphingosine (d17:1), Avanti Research - A Croda Brand 860640P, powder
Avanti
17:0-17:1-17:0 D5 TG, Avanti Research - A Croda Brand 860903P, powder
Avanti
18:1-2:0 DG, 1-oleoyl-2-acetyl-sn-glycerol, neat oil
Avanti
12:0 DG, Avanti Research - A Croda Brand
Avanti
12:0-13:0 PC, Avanti Research - A Croda Brand LM1000, methanol solution
Avanti
18:1-2:0 DG, 1-oleoyl-2-acetyl-sn-glycerol, chloroform