Merck
  • Serum Amino Acid Profiles in Normal Subjects and in Patients with or at Risk of Alzheimer Dementia.
Serum Amino Acid Profiles in Normal Subjects and in Patients with or at Risk of Alzheimer Dementia.
Dementia and geriatric cognitive disorders extra (2017-06-20)
Gaetano Corso, Adriana Cristofano, Nadia Sapere, Giancarlo la Marca, Antonella Angiolillo, Michela Vitale, Roberto Fratangelo, Teresa Lombardi, Carola Porcile, Mariano Intrieri, Alfonso Di Costanzo
ABSTRACT
Abnormalities in the plasma amino acid profile have been reported in Alzheimer disease (AD), but no data exist for the prodromal phase characterized by subjective memory complaint (SMC). It was our aim to understand if serum amino acid levels change along the continuum from normal to AD, and to identify possible diagnostic biomarkers. Serum levels of 15 amino acids and 2 organic acids were determined in 4 groups of participants - 29 with probable AD, 18 with mild cognitive impairment (MCI), 24 with SMC, and 46 cognitively healthy subjects (HS) - by electrospray tandem mass spectrometry. Glutamate, aspartate, and phenylalanine progressively decreased, while citrulline, argi-ninosuccinate, and homocitrulline progressively increased, from HS over SMC and MCI to AD. The panel including these 6 amino acids and 4 ratios (glutamate/citrulline, citrulline/phenylalanine, leucine plus isoleucine/phenylalanine, and arginine/phenylalanine) discriminated AD from HS with about 96% accuracy. Other panels including 20 biomarkers discriminated SMC or MCI from AD or HS with an accuracy ranging from 88 to 75%. Amino acids contribute to a characteristic metabotype during the progression of AD along the continuum from health to frank dementia, and their monitoring in elderly individuals might help to detect at-risk subjects.
Sigma-Aldrich
L-Homocitrulline, ≥95% (HPLC)