Merck
  • The mutation m.13513G>A impairs cardiac function, favoring a neuroectoderm commitment, in a mutant-load dependent way.
The mutation m.13513G>A impairs cardiac function, favoring a neuroectoderm commitment, in a mutant-load dependent way.
Journal of cellular physiology (2019-04-06)
Teresa Galera-Monge, Francisco Zurita-Díaz, Rafael Garesse, María Esther Gallardo
ABSTRACT
Mitochondrial disorders (MDs) arise as a result of a respiratory chain dysfunction. While some MDs can affect a single organ, many involve several organs, the brain being the most affected, followed by heart and/or muscle. Many of these diseases are associated with heteroplasmic mutations in the mitochondrial DNA (mtDNA). The proportion of mutated mtDNA must exceed a critical threshold to produce disease. Therefore, understanding how embryonic development determines the heteroplasmy level in each tissue could explain the organ susceptibility and the clinical heterogeneity observed in these patients. In this report, the dynamics of heteroplasmy and the influence in cardiac commitment of the mutational load of the m.13513G>A mutation has been analyzed. This mutation has been reported as a frequent cause of Leigh syndrome (LS) and is commonly associated with cardiac problems. In this report, induced pluripotent stem cell (iPSc) technology has been used to delve into the molecular mechanisms underlying cardiac disease in LS. When mutation m.13513G>A is above a threshold, iPSc-derived cardiomyocytes (iPSc-CMs) could not be obtained due to an inefficient epithelial-mesenchymal transition. Surprisingly, these cells are redirected toward neuroectodermal lineages that would give rise to the brain. However, when mutation is below that threshold, dysfunctional CM are generated in a mutant-load dependent way. We suggest that distribution of the m.13513G>A mutation during cardiac differentiation is not at random. We propose a possible explanation of why neuropathology is a frequent feature of MD, but cardiac involvement is not always present.
Sigma-Aldrich
Alkaline Phosphatase Blue Membrane Substrate Solution, sufficient for 400 mL working substrate
Sigma-Aldrich
Monoclonal Anti-AFP antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-TNNT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse