Merck
  • Replication Fork Activation Is Enabled by a Single-Stranded DNA Gate in CMG Helicase.
Replication Fork Activation Is Enabled by a Single-Stranded DNA Gate in CMG Helicase.
Cell (2019-07-28)
Michael R Wasserman, Grant D Schauer, Michael E O'Donnell, Shixin Liu
ABSTRACT
The eukaryotic replicative helicase CMG is a closed ring around double-stranded (ds)DNA at origins yet must transition to single-stranded (ss)DNA for helicase action. CMG must also handle repair intermediates, such as reversed forks that lack ssDNA. Here, using correlative single-molecule fluorescence and force microscopy, we show that CMG harbors a ssDNA gate that enables transitions between ss and dsDNA. When coupled to DNA polymerase, CMG remains on ssDNA, but when uncoupled, CMG employs this gate to traverse forked junctions onto dsDNA. Surprisingly, CMG undergoes rapid diffusion on dsDNA and can transition back onto ssDNA to nucleate a functional replisome. The gate-distinct from that between Mcm2/5 used for origin loading-is intrinsic to CMG; however, Mcm10 promotes strand passage by enhancing the affinity of CMG to DNA. This gating process may explain the dsDNA-to-ssDNA transition of CMG at origins and help preserve CMG on dsDNA during fork repair.
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
N-Bromoacetamide, powder
Millipore
Middlebrook 7H10 Agar Base, suitable for microbiology, NutriSelect® Plus, for use in isolation and cultivation of Mycobacterium species
Roche
DNA, lambda, from bacteriophage lambda cl857 Sam 7
Sigma-Aldrich
DBCO-Cy5, for Copper-free Click Chemistry
Sigma-Aldrich
Cyclooctatetraene, 98%
Sigma-Aldrich
4-Nitrobenzyl alcohol, 99%
Sigma-Aldrich
Coenzyme A trilithium salt, ≥93%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
BLR(DE3) Competent Cells - Novagen, BLR is a recA derivative of BL21 that improves plasmid monomer yields and may help stabilize target plasmids containing repetitive sequences.