Merck
  • Retention time profiling of all 136 tetra- through octa- chlorinated dibenzo-p-dioxins and dibenzofurans on a variety of Si-Arylene gas chromatographic stationary phases.
Retention time profiling of all 136 tetra- through octa- chlorinated dibenzo-p-dioxins and dibenzofurans on a variety of Si-Arylene gas chromatographic stationary phases.
Chemosphere (2011-07-09)
Vyacheslav N Fishman, Gregory D Martin, Michael Wilken
ABSTRACT
The applicability of three different Silphenylene Silicone co-polymer (Si-Arylene) GC stationary phases (J&W Scientific DB-5ms, Varian VF-5ms, and VF-Xms) has been evaluated for the separation of all 136 tetra- through the octa- chlorinated dibenzo-p-dioxins (PCDD) and chlorinated dibenzofurans (PCDF) from closely eluting isomers using gas chromatography/high resolution mass spectrometry (GC/HRMS). Their relative performance data are compared to the "conventional" 5% diphenyl 95% dimethylpolysiloxane GC column (Supelco Equity-5) and to each other based on absolute retention times, visualized mass chromatograms, and the separation of 2,3,7,8-substituted isomers. VF-Xms GC column was able to demonstrate a better performance towards separation of 2,3,7,8-substituted PCDD/PCDF compared to other Si-Arylene GC columns tested, where only 2,3,4,7,8-PnCDF can not be resolved from 1,2,3,6,9-PnCDF and 2,3,4,6,7,8-/1,2,3,6,8,9-HxCDF and 1,2,3,7,8,9-/1,2,3,4,8,9-HxCDF can be at least partially resolved. These data suggest that the development of a single GC column for the separation of all 17 2,3,7,8-substituted dioxins and furans is feasible, which therefore could be used as a standard tool for the PCDD/PCDF methods globally including USEPA methods 1613b, 8290A, 8280B, European Standard Method EN 1948, Canadian methods DFPCB-E3418, EPS 1/RM/19, Japanese methods JIS K0311 and JIS K0312. Having one specific GC column to separate all 2,3,7,8-substituted PCDD/PCDF will significantly improve the data quality, comparability by the various methods and assessment techniques while simultaneously leading to a more cost and time efficient operation.
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 50 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 500 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 1,000 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 5 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 200 cSt (25 °C)(lit.)