Merck
  • Cellular inhibitors of apoptosis proteins cIAP1 and cIAP2 are required for efficient caspase-1 activation by the inflammasome.
Cellular inhibitors of apoptosis proteins cIAP1 and cIAP2 are required for efficient caspase-1 activation by the inflammasome.
Immunity (2011-12-27)
Katherine Labbé, Christian R McIntire, Karine Doiron, Philippe M Leblanc, Maya Saleh
ABSTRACT
Pathogen and danger recognition by the inflammasome activates inflammatory caspases that mediate inflammation and cell death. The cellular inhibitor of apoptosis proteins (cIAPs) function in apoptosis and innate immunity, but their role in modulating the inflammasome and the inflammatory caspases is unknown. Here we report that the cIAPs are critical effectors of the inflammasome and are required for efficient caspase-1 activation. cIAP1, cIAP2, and the adaptor protein TRAF2 interacted with caspase-1-containing complexes and mediated the activating nondegradative K63-linked polyubiquitination of caspase-1. Deficiency in cIAP1 (encoded by Birc2) or cIAP2 (Birc3) impaired caspase-1 activation after spontaneous or agonist-induced inflammasome assembly, and Birc2(-/-) or Birc3(-/-) mice or mice administered with an IAP antagonist had a dampened response to inflammasome agonists and were resistant to peritonitis. Our results describe a role for the cIAPs in innate immunity and further demonstrate the evolutionary conservation between cell death and inflammation mechanisms.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Millipore
ProteinA-Sepharose 4, Fast Flow from Staphylococcus aureus, aqueous ethanol suspension
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone HWA4C4, clone HWA4C4, from mouse
Millipore
Protein G Sepharose, Fast Flow, recombinant, expressed in E. coli, aqueous ethanol suspension