Merck
  • Cystatin C as a p53-inducible apoptotic mediator that regulates cathepsin L activity.
Cystatin C as a p53-inducible apoptotic mediator that regulates cathepsin L activity.
Cancer science (2016-01-13)
Jinichi Mori, Chizu Tanikawa, Yuki Funauchi, Paulisally Hau Yi Lo, Yusuke Nakamura, Koichi Matsuda
ABSTRACT
In response to various cellular stresses, p53 is activated and inhibits malignant transformation through the transcriptional regulation of its target genes. However, the full picture of the p53 downstream pathway still remains to be elucidated. Here we identified cystatin C, a major inhibitor of cathepsins, as a novel p53 target. In response to DNA damage, activated p53 induced cystatin C expression through p53 binding sequence in the first intron. We showed that cathepsin L activity was decreased in HCT116 p53(+/+) cells after adriamycin treatment, but not in HCT116 p53(-/-) cells. We also found that knockdown of cystatin C reduced adriamycin-induced caspase-3 activation. Cystatin C expression was significantly downregulated in breast cancer cells with p53 mutations, and decreased cystatin C expression was associated with poor prognosis of breast cancer. Our findings revealed an important role of the p53-cystatin C pathway in human carcinogenesis.
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Sigma-Aldrich
Anti-p53 (Ab-12) (Pantropic) Mouse mAb (DO-7), liquid, clone DO-7, Calbiochem®