Merck
  • sn-1,2-Dioctanoylglycerol. A cell-permeable diacylglycerol that mimics phorbol diester action on the epidermal growth factor receptor and mitogenesis.
sn-1,2-Dioctanoylglycerol. A cell-permeable diacylglycerol that mimics phorbol diester action on the epidermal growth factor receptor and mitogenesis.
The Journal of biological chemistry (1985-02-10)
R J Davis, B R Ganong, R M Bell, M P Czech
ABSTRACT
The cell-permeable diacylglycerol, sn-1,2-dioctanoylglycerol (DiC8), is shown to mimic the effect of tumor promoting phorbol diesters on epidermal growth factor (EGF) binding and action in intact cells. DiC8 inhibited the binding of [3H]phorbol dibutyrate to A431 cell monolayers indicating that the diacylglycerol interacts with the phorbol diester receptor. At 0.3 microM, DiC8 half-maximally inhibited the high affinity binding of 125I-EGF to A431 human epidermoid carcinoma cells. Scatchard analysis indicated that the inhibition of 125I-EGF binding was very similar to that observed in the presence of 4 beta-phorbol 12 beta-myristate 13 alpha-acetate (PMA). DiC8 also mimicked the action of PMA to increase the phosphorylation state of the EGF receptor in 32P-labeled cells. Phosphoamino acid analysis demonstrated that DiC8 and PMA caused an increase in the level of EGF-receptor phosphoserine and phosphothreonine, whereas EGF caused an increase in the level of phosphoserine, phosphothreonine, and phosphotyrosine. Phosphopeptide mapping of the EGF receptor showed that DiC8 and PMA enhanced the phosphorylation of the same tryptic peptides. DiC8 inhibited the EGF-dependent tyrosine phosphorylation of the EGF receptor in A431 cells in a similar manner to that observed with PMA. In further experiments with quiescent Swiss 3T3 fibroblasts, DiC8 mimicked the ability of PMA to stimulate the incorporation of [methyl-3H]thymidine synergistically with low concentrations of EGF. This result indicates that DiC8 will mimic the long-term effects of PMA to regulate mitogenesis and raises the possibility that it may be active in two stage carcinogenesis. As both DiC8 and PMA stimulate the Ca2+- and phospholipid-dependent protein kinase (C-kinase) in vitro, the results support the hypothesis that the activation of C-kinase is a critical component of phorbol diester action on EGF receptor modulation and cell proliferation.
Product Description
Avanti
16:0 DG, 1,2-dipalmitoyl-sn-glycerol, chloroform
Avanti
16:0 DG, 1,2-dipalmitoyl-sn-glycerol, powder
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, neat oil
Avanti
10:0 DG, 1,2-didecanoyl-sn-glycerol, powder
Avanti
18:0-18:2 DG, 1-stearoyl-2-linoleoyl-sn-glycerol, chloroform
Avanti
18:1 DG, Avanti Research - A Croda Brand
Avanti
18:1 DG, 1-2-dioleoyl-sn-glycerol, chloroform
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, chloroform solution
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, chloroform
Avanti
18:0-20:4 DG, 1-stearoyl-2-arachidonoyl-sn-glycerol, chloroform
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, neat oil
Avanti
14:0 DG, 1,2-dimyristoyl-sn-glycerol, powder
Avanti
18:0-22:6 DG, 1-stearoyl-2-docosahexaenoyl-sn-glycerol, chloroform
Avanti
12:0 DG, Avanti Research - A Croda Brand
Avanti
18:0 DG, 1,2-dioctadecanoyl-sn-glycerol, powder
Avanti
18:1-2:0 DG, 1-oleoyl-2-acetyl-sn-glycerol, chloroform
Avanti
18:1-2:0 DG, 1-oleoyl-2-acetyl-sn-glycerol, neat oil