Merck
Nuclear cGAS suppresses DNA repair and promotes tumorigenesis.
Nature (2018-10-26)
Haipeng Liu, Haiping Zhang, Xiangyang Wu, Dapeng Ma, Juehui Wu, Lin Wang, Yan Jiang, Yiyan Fei, Chenggang Zhu, Rong Tan, Peter Jungblut, Gang Pei, Anca Dorhoi, Qiaoling Yan, Fan Zhang, Ruijuan Zheng, Siyu Liu, Haijiao Liang, Zhonghua Liu, Hua Yang, Jianxia Chen, Peng Wang, Tianqi Tang, Wenxia Peng, Zhangsen Hu, Zhu Xu, Xiaochen Huang, Jie Wang, Haohao Li, Yilong Zhou, Feng Liu, Dapeng Yan, Stefan H E Kaufmann, Chang Chen, Zhiyong Mao, Baoxue Ge
ABSTRACT
Accurate repair of DNA double-stranded breaks by homologous recombination preserves genome integrity and inhibits tumorigenesis. Cyclic GMP-AMP synthase (cGAS) is a cytosolic DNA sensor that activates innate immunity by initiating the STING-IRF3-type I IFN signalling cascade1,2. Recognition of ruptured micronuclei by cGAS links genome instability to the innate immune response3,4, but the potential involvement of cGAS in DNA repair remains unknown. Here we demonstrate that cGAS inhibits homologous recombination in mouse and human models. DNA damage induces nuclear translocation of cGAS in a manner that is dependent on importin-α, and the phosphorylation of cGAS at tyrosine 215-mediated by B-lymphoid tyrosine kinase-facilitates the cytosolic retention of cGAS. In the nucleus, cGAS is recruited to double-stranded breaks and interacts with PARP1 via poly(ADP-ribose). The cGAS-PARP1 interaction impedes the formation of the PARP1-Timeless complex, and thereby suppresses homologous recombination. We show that knockdown of cGAS suppresses DNA damage and inhibits tumour growth both in vitro and in vivo. We conclude that nuclear cGAS suppresses homologous-recombination-mediated repair and promotes tumour growth, and that cGAS therefore represents a potential target for cancer prevention and therapy.
Sigma-Aldrich
PARP1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Supelco
o-Tolidine, analytical standard
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
PARP-1 human, recombinant, expressed in E. coli