Merck
  • Synthesis of the cyclohexane core of phomactins and a new route to the bicyclo[9.3.1]pentadecane diterpenoid skeleton.
Synthesis of the cyclohexane core of phomactins and a new route to the bicyclo[9.3.1]pentadecane diterpenoid skeleton.
Organic letters (2010-12-18)
Keith D Schwartz, James D White
ABSTRACT
Conjugate reduction of an enone accompanied by in situ intramolecular aldol condensation was used to construct the tetrasubstituted cyclohexane nucleus of phomactins. Subsequent relay ring-closing metathesis completed the nine-membered ansa bridge of the diterpenoid framework.
Product Description
Supelco
Pentadecane, analytical standard
Sigma-Aldrich
Pentadecane, ≥99%
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)