Merck
  • Non-canonical Activation of the DNA Sensing Adaptor STING by ATM and IFI16 Mediates NF-κB Signaling after Nuclear DNA Damage.
Non-canonical Activation of the DNA Sensing Adaptor STING by ATM and IFI16 Mediates NF-κB Signaling after Nuclear DNA Damage.
Molecular cell (2018-09-08)
Gillian Dunphy, Sinéad M Flannery, Jessica F Almine, Dympna J Connolly, Christina Paulus, Kasper L Jønsson, Martin R Jakobsen, Michael M Nevels, Andrew G Bowie, Leonie Unterholzner
ABSTRACT
DNA damage can be sensed as a danger-associated molecular pattern by the innate immune system. Here we find that keratinocytes and other human cells mount an innate immune response within hours of etoposide-induced DNA damage, which involves the DNA sensing adaptor STING but is independent of the cytosolic DNA receptor cGAS. This non-canonical activation of STING is mediated by the DNA binding protein IFI16, together with the DNA damage response factors ATM and PARP-1, resulting in the assembly of an alternative STING signaling complex that includes the tumor suppressor p53 and the E3 ubiquitin ligase TRAF6. TRAF6 catalyzes the formation of K63-linked ubiquitin chains on STING, leading to the activation of the transcription factor NF-κB and the induction of an alternative STING-dependent gene expression program. We propose that STING acts as a signaling hub that coordinates a transcriptional response depending on its mode of activation.
Sigma-Aldrich
Anti-MB21D1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Polyinosinic–polycytidylic acid potassium salt, with buffer salts, TLR ligand tested
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
NovaBlue(DE3) Competent Cells - Novagen, NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, DE3 lysogen cannot be used for the blue/white screening of recombinant plasmids
Sigma-Aldrich
Giemsa Stain, Modified
Sigma-Aldrich
Anti-acetyl-Histone H3 (Ac-Lys9) antibody produced in rabbit, 2-2.5 mg/mL, affinity isolated antibody, buffered aqueous solution