Merck
  • The Spastic Paraplegia-Associated Phospholipase DDHD1 Is a Primary Brain Phosphatidylinositol Lipase.
The Spastic Paraplegia-Associated Phospholipase DDHD1 Is a Primary Brain Phosphatidylinositol Lipase.
Biochemistry (2018-09-18)
Jordon M Inloes, Hui Jing, Benjamin F Cravatt
ABSTRACT
Deleterious mutations in the serine hydrolase DDHD domain containing 1 (DDHD1) cause the SPG28 subtype of the neurological disease hereditary spastic paraplegia (HSP), which is characterized by axonal neuropathy and gait impairments. DDHD1 has been shown to display PLA1-type phospholipase activity with a preference for phosphatidic acid. However, the endogenous lipid pathways regulated by DDHD1 in vivo remain poorly understood. Here we use a combination of untargeted and targeted metabolomics to compare the lipid content of brain tissue from DDHD1+/+ and DDHD1-/- mice, revealing that DDHD1 inactivation causes a substantial decrease in the level of polyunsaturated lysophosphatidylinositol (LPI) lipids and a corresponding increase in the level of phosphatidylinositol (PI) lipids. Levels of other phospholipids were mostly unchanged, with the exception of decreases in the levels of select polyunsaturated lysophosphatidylserine (LPS) and lysophosphatidylcholine lipids and a striking remodeling of PI phosphates (e.g., PIP and PIP2) in DDHD1-/- brain tissue. Biochemical assays confirmed that DDHD1 hydrolyzes PI/PS to LPI/LPS with sn-1 selectivity and accounts for a substantial fraction of the PI/PS lipase activity in mouse brain tissue. These data indicate that DDHD1 is a principal regulator of bioactive LPI and other lysophospholipids, as well as PI phosphates, in the mammalian nervous system, pointing to a potential role for these lipid pathways in HSP.
Product Description
Avanti
16:0-d31-18:1 PE, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
16:0-d31-18:1 PG, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-[phospho-rac-(1-glycerol)] (sodium salt), chloroform
Avanti
16:0-d31-18:1 PE, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
16:0-d31-18:1 PG, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-[phospho-rac-(1-glycerol)] (sodium salt), powder
Avanti
12:0 PE, 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
17:1 Lyso PI, 1-(10Z-heptadecenoyl)-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt), powder
Avanti
14:0 BMP (S,R), bis(monomyristoylglycero)phosphate (S,R Isomer) (ammonium salt), powder
Avanti
17:1 Lyso PE, 1-(10Z-heptadecenoyl)-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
17:0 PS, Avanti Research - A Croda Brand
Avanti
17:0-20:4 PI(4,5)P2, Avanti Research - A Croda Brand LM1904, powder
Avanti
17:0-20:4 PI(3)P, Avanti Research - A Croda Brand
Avanti
12:0 PC (DLPC), Avanti Research - A Croda Brand
Avanti
17:0 Lyso PC, 1-heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
17:0 Lyso PC, Avanti Research - A Croda Brand
Avanti
17:1 Lyso PS, 1-(10Z-heptadecenoyl)-2-hydroxy-sn-glycero-3-[phospho-L-serine] (sodium salt), chloroform
Avanti
12:0 PC (DLPC), 1,2-dilauroyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-d31-18:1 PA, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
17:1 Lyso PS, 1-(10Z-heptadecenoyl)-2-hydroxy-sn-glycero-3-[phospho-L-serine] (sodium salt), powder
Avanti
16:0-d31-18:1 PI, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol (ammonium salt), powder
Avanti
16:0-d31-18:1 PC, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-d31-18:1 PA, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphate (sodium salt), powder
Avanti
16:0-d31-18:1 PC, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, powder
Avanti
17:1 Lyso PG, 1-(10Z-heptadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform