Merck
  • Critical role of truncated α-synuclein and aggregates in Parkinson's disease and incidental Lewy body disease.
Critical role of truncated α-synuclein and aggregates in Parkinson's disease and incidental Lewy body disease.
Brain pathology (Zurich, Switzerland) (2012-03-29)
Kavita Prasad, Thomas G Beach, John Hedreen, Eric K Richfield
ABSTRACT
The role of Lewy bodies, Lewy neurites and α-synuclein (αSYN) in the pathophysiology and diagnosis of Parkinson's disease (PD) is unclear. We used postmortem human tissue, a panel of antibodies (Abs) and confocal microscopy to examine the three-dimensional neurochemical anatomy of the nigrostriatal system. Abs were specific to truncated (tαSYN), phosphorylated and full-length αSYN. The findings demonstrate the critical role of tαSYN in initiating aggregation, a role for other forms of αSYN in aggregate expansion, a reason for the wide variety of proteins present in different aggregates, an explanation for the laminar appearance of aggregates described historically using different methods, the existence of proximal greater than distal aggregation in the vulnerable nigrostriatal pathway, the independent transport of different forms of αSYN as cargo along axons and a possible sequence for the formation of Lewy bodies. Findings differed between incidental Lewy body disease and PD only quantitatively. These findings have implications for understanding the pathogenesis and treatment of PD.
Sigma-Aldrich
Anti-α-Synuclein antibody, Mouse monoclonal, clone Syn211, purified from hybridoma cell culture
Pricing and availability is not currently available.