Merck
  • The formation of ceramide-1-phosphate during neutrophil phagocytosis and its role in liposome fusion.
The formation of ceramide-1-phosphate during neutrophil phagocytosis and its role in liposome fusion.
The Journal of biological chemistry (1998-12-05)
V T Hinkovska-Galcheva, L A Boxer, P J Mansfield, D Harsh, A Blackwood, J A Shayman
ABSTRACT
Ceramide, a product of agonist-stimulated sphingomyelinase activation, is known to be generated during the phagocytosis of antibody-coated erythrocytes by polymorphonuclear leukocytes. Agonist-stimulated formation of ceramide-1-phosphate is now shown to occur in 32PO4-labeled neutrophils. Ceramide-1-phosphate is formed by a calcium-dependent ceramide kinase, found predominately in the neutrophil plasma membrane. The neutrophil kinase is specific for ceramide because, in contrast to the bacterial diglyceride kinase, ceramide is not phosphorylated under conditions specific for diglyceride phosphorylation. Conversely, 1,2-diacylglycerol does not serve as substrate for the neutrophil ceramide kinase. Ceramide kinase activation occurs in a time-dependent fashion, reaching peak activity 10 min after formyl peptide stimulation and challenge with antibody-coated erythrocytes. The lipid kinase activity is optimal at pH 6.8. Because the formation of the phagolysosome is a critical event in phagocytosis, the effect of ceramide-1-phosphate in promoting the fusion of liposomes was determined. Both the addition of increasing concentrations of sphingomyelinase D and ceramide-1-phosphate promoted liposomal fusion. In summary, ceramide-1-phosphate is formed during phagocytosis through activation of ceramide kinase. Ceramide-1-phosphate may promote phagolysosome formation.
Product Description
Avanti
C16 Ceramide-1-Phosphate (d18:1/16:0), Avanti Research - A Croda Brand 860533P, powder
Avanti
C12 Ceramide-1-Phosphate (d18:1/12:0), Avanti Research - A Croda Brand 860531P, powder
Avanti
C8 Ceramide-1-Phosphate (d18:1/8:0), N-octanoyl-ceramide-1-phosphate (ammonium salt), powder
Avanti
C24 Ceramide-1-Phosphate (d18:1/24:0), Avanti Research - A Croda Brand 860527P, powder
Avanti
C8 Ceramide-1-Phosphate (d17:1/8:0), Avanti Research - A Croda Brand 860652P, powder
Avanti
C2 Ceramide-1-Phosphate (d18:1/2:0), N-acetoyl-ceramide-1-phosphate (ammonium salt), powder