Merck
  • Mild electrophilic trifluoromethylation of secondary and primary aryl- and alkylphosphines using hypervalent iodine(iii)-CF(3) reagents.
Mild electrophilic trifluoromethylation of secondary and primary aryl- and alkylphosphines using hypervalent iodine(iii)-CF(3) reagents.
Chemical communications (Cambridge, England) (2008-03-21)
Patrick Eisenberger, Iris Kieltsch, Nicolas Armanino, Antonio Togni
ABSTRACT
A direct, mild and efficient trifluoromethylation of primary and secondary phosphines is achieved with easily accessible, cheap hypervalent iodine compounds acting as electrophilic CF(3)-transfer reagents.
Sigma-Aldrich
3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%
Pricing and availability is not currently available.