Merck
  • Triggering the interferon antiviral response through an IKK-related pathway.
Triggering the interferon antiviral response through an IKK-related pathway.
Science (New York, N.Y.) (2003-04-19)
Sonia Sharma, Benjamin R tenOever, Nathalie Grandvaux, Guo-Ping Zhou, Rongtuan Lin, John Hiscott
ABSTRACT
Rapid induction of type I interferon expression, a central event in establishing the innate antiviral response, requires cooperative activation of numerous transcription factors. Although signaling pathways that activate the transcription factors nuclear factor kappaB and ATF-2/c-Jun have been well characterized, activation of the interferon regulatory factors IRF-3 and IRF-7 has remained a critical missing link in understanding interferon signaling. We report here that the IkappaB kinase (IKK)-related kinases IKKepsilon and TANK-binding kinase 1 are components of the virus-activated kinase that phosphorylate IRF-3 and IRF-7. These studies illustrate an essential role for an IKK-related kinase pathway in triggering the host antiviral response to viral infection.
Sigma-Aldrich
TBK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.