Merck
Phagocytosis-mediated M1 activation by chitin but not by chitosan.
American journal of physiology. Cell physiology (2018-05-03)
Spring Davis, Aiko M Cirone, Janet Menzie, Floyd Russell, C Kathleen Dorey, Yoshimi Shibata, Jianning Wei, Changlong Nan
ABSTRACT
Chitin particles have been used to understand host response to chitin-containing pathogens and allergens and are known to induce a wide range of polarized macrophage activations, depending, at least in part, on particle size. Nonphagocytosable particles larger than a macrophage induce tissue repair M2 activation. In contrast, phagocytosable chitin microparticles (CMPs, 1-10 μm diameters) induce M1 macrophages that kill intracellular microbes and damage tissues. However, chitosan (deacetylated) microparticles (de-CMPs, 1-10 µm) induce poor M1 activation. Toll-like receptor 2 (TLR2) and associated coreceptors in macrophages appear to be required for the M1 activation. To understand the exact mechanism of phagocytosis-mediated M1 activation by chitin, we isolated macrophage proteins that bind to CMPs during early phagocytosis and determined that TLR1, TLR2, CD14, late endosomal/lysosomal adaptor MAPK and mechanistic target of rapamycin activator 1 (LAMTOR1), Lck/Yes novel tyrosine kinase (Lyn), and β-actin formed phagosomal CMP-TLR2 clusters. These proteins were also detected in TLR2 phagosomal clusters in macrophages phagocytosing de-CMPs, but at relatively lower levels than in the CMP-TLR2 clusters. Importantly, CMP-TLR2 clusters further recruited myeloid differentiation primary response gene 88 (MyD88) and Toll-IL-1 receptor-containing adaptor protein (TIRAP) and phosphorylated Lyn, whereas neither the adaptors nor phosphorylated Lyn was detected in the de-CMP clusters. The results indicate that the acetyl group played an obligatory, phagocytosis-dependent role in the initiation of an integrated signal for TLR2-mediated M1 activation.
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit