Merck
  • Autophagy-mediated neuroprotection induced by octreotide in an ex vivo model of early diabetic retinopathy.
Autophagy-mediated neuroprotection induced by octreotide in an ex vivo model of early diabetic retinopathy.
Pharmacological research (2017-10-04)
Rosario Amato, Elisabetta Catalani, Massimo Dal Monte, Maurizio Cammalleri, Ilaria Di Renzo, Cristiana Perrotta, Davide Cervia, Giovanni Casini
ABSTRACT
Neuronal injury plays a major role in diabetic retinopathy (DR). Our hypothesis was that the balance between neuronal death and survival may depend on a similar equilibrium between apoptosis and autophagy and that a neuroprotectant may act by influencing this equilibrium. Ex vivo mouse retinal explants were treated with high glucose (HG) for 10days and the somatostatin analog octreotide (OCT) was used as a neuroprotectant. Chloroquine (CQ) was used as an autophagy inhibitor. Apoptotic and autophagic markers were evaluated using western blot and immunohistochemistry. HG-treated explants displayed a significant increase of apoptosis paralleled by a significant decrease of the autophagic flux, which was likely to be due to increased activity of the autophagy regulator mTOR (mammalian target of rapamycin). Treatment with OCT rescued HG-treated retinal explants from apoptosis and determined an increase of autophagic activity with concomitant mTOR inhibition. Blocking the autophagic flux with CQ completely abolished the anti-apoptotic effect of OCT. Immunohistochemical observations showed that OCT-induced autophagy is localized to populations of bipolar and amacrine cells and to ganglion cells. These observations revealed the antithetic role of apoptosis and autophagy, highlighting their equilibrium from which neuronal survival is likely to depend. These data suggest the crucial role covered by autophagy, which could be considered as a molecular target for DR neuroprotective treatment strategies.
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-p62/SQSTM1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GABA Transporter-1 Antibody, CT (a.a. 588-599), Chemicon®, from rabbit
Sigma-Aldrich
Octreotide, ≥98% (HPLC)
Sigma-Aldrich
Anti-Caspase 3, Active antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-DAB1 antibody produced in rabbit, affinity isolated antibody