Merck
  • Survey of differentially expressed proteins and genes in jasmonic acid treated rice seedling shoot and root at the proteomics and transcriptomics levels.
Survey of differentially expressed proteins and genes in jasmonic acid treated rice seedling shoot and root at the proteomics and transcriptomics levels.
Journal of proteome research (2007-08-23)
Kyoungwon Cho, Ganesh Kumar Agrawal, Junko Shibato, Young-Ho Jung, Yeon-Ki Kim, Baek Hie Nahm, Nam-Soo Jwa, Shigeru Tamogami, Oksoo Han, Kimiyoshi Kohda, Hitoshi Iwahashi, Randeep Rakwal
ABSTRACT
Two global approaches were applied to develop an inventory of differentially expressed proteins and genes in rice (cv. Nipponbare) seedling grown on Murashige and Skoog medium with and without jasmonic acid (JA). JA significantly reduced the growth of shoot, root, leaf, and leaf sheath depending on JA concentration (1, 2, 5, 10, 25, and 50 microM) as compared with control. Almost 50% growth inhibition of seedling was observed with 5 microM JA. Shoots and roots of seedlings grown on 5 microM JA for 7 days were then used for proteomics and transcriptomics analyses. Two-dimensional gel electrophoresis revealed 66 and 68 differentially expressed protein spots in shoot and root, respectively, compared to their respective controls. Tandem mass spectrometry analysis of these proteins identified 52 (shoot) and 56 (root) nonredundant proteins, belonging to 10 functional categories. Proteins involved in photosynthesis (44%), cellular respiratory (11%), and protein modification and chaperone (11%) were highly represented in shoot, whereas antioxidant system (18%), cellular respiratory (17%), and defense-related proteins (15%) were highly represented in root. Transcriptomics analysis of shoot and root identified 107 and 325 induced genes and 34 and 213 suppressed genes in shoot and root, respectively. Except of unknown genes with over 57% of the total, most genes encode for proteins involved in secondary metabolism, energy production, protein modification and chaperone, transporters, and cytochrome P450. These identified proteins and genes have been discussed with respect to the JA-induced phenotype providing a new insight into the role of JA in rice seedling growth and development. phytohormone * inhibitory concentration * growth * gel-based approach * mass spectrometry * DNA microarray.
Sigma-Aldrich
(±)-Jasmonic acid, BioReagent, suitable for plant cell culture, liquid