Merck
  • Fucosylated Molecules Competitively Interfere with Cholera Toxin Binding to Host Cells.
Fucosylated Molecules Competitively Interfere with Cholera Toxin Binding to Host Cells.
ACS infectious diseases (2018-02-08)
Amberlyn M Wands, Jakob Cervin, He Huang, Ye Zhang, Gyusaang Youn, Chad A Brautigam, Maria Matson Dzebo, Per Björklund, Ville Wallenius, Danielle K Bright, Clay S Bennett, Pernilla Wittung-Stafshede, Nicole S Sampson, Ulf Yrlid, Jennifer J Kohler
ABSTRACT
Cholera toxin (CT) enters host intestinal epithelia cells, and its retrograde transport to the cytosol results in the massive loss of fluids and electrolytes associated with severe dehydration. To initiate this intoxication process, the B subunit of CT (CTB) first binds to a cell surface receptor displayed on the apical surface of the intestinal epithelia. While the monosialoganglioside GM1 is widely accepted to be the sole receptor for CT, intestinal epithelial cell lines also utilize fucosylated glycan epitopes on glycoproteins to facilitate cell surface binding and endocytic uptake of the toxin. Further, l-fucose can competively inhibit CTB binding to intestinal epithelia cells. Here, we use competition binding assays with l-fucose analogs to decipher the molecular determinants for l-fucose inhibition of cholera toxin subunit B (CTB) binding. Additionally, we find that mono- and difucosylated oligosaccharides are more potent inhibitors than l-fucose alone, with the LeY tetrasaccharide emerging as the most potent inhibitor of CTB binding to two colonic epithelial cell lines (T84 and Colo205). Finally, a non-natural fucose-containing polymer inhibits CTB binding two orders of magnitude more potently than the LeY glycan when tested against Colo205 cells. This same polymer also inhibits CTB binding to T84 cells and primary human jejunal epithelial cells in a dose-dependent manner. These findings suggest the possibility that polymeric display of fucose might be exploited as a prophylactic or therapeutic approach to block the action of CT toward the human intestinal epithelium.
Sigma-Aldrich
CTB, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
L-(−)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
N-Acetylneuraminic acid, ≥98% (HPLC), from Escherichia coli
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acetoxyacetic acid, 99%
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
3′-Sialyllactose, from bovine milk or colostrum, ≥97% (HPLC)
Sigma-Aldrich
5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride, ≥90% (HPLC)