Merck
  • Determination of steroid hormones and their metabolite in several types of meat samples by ultra high performance liquid chromatography-Orbitrap high resolution mass spectrometry.
Determination of steroid hormones and their metabolite in several types of meat samples by ultra high performance liquid chromatography-Orbitrap high resolution mass spectrometry.
Journal of chromatography. A (2018-02-06)
Marina López-García, Roberto Romero-González, Antonia Garrido Frenich
ABSTRACT
A new analytical method based on ultra-high performance liquid chromatography (UHPLC) coupled to Orbitrap high resolution mass spectrometry (Orbitrap-HRMS) has been developed for the determination of steroid hormones (hydrocortisone, cortisone, progesterone, prednisone, prednisolone, testosterone, melengesterol acetate, hydrocortisone-21-acetate, cortisone-21-acetate, testosterone propionate, 17α-methyltestosterone, 6α-methylprednisolone and medroxyprogesterone) and their metabolite (17α-hydroxyprogesterone) in three meat samples (chicken, pork and beef). Two different extraction approaches were tested (QuEChERS "quick, easy, cheap, effective, rugged and safe" and "dilute and shoot"), observing that the QuEChERS method provided the best results in terms of recovery. A clean-up step was applied comparing several sorbents, obtaining the best results when florisil and aluminum oxide were used. The optimized method was validated, obtaining suitable results for all validation parameters in the three meat matrices evaluated. Recovery values ranged from 70% to 103% (except for prednisone in beef samples), meanwhile repeatability and reproducibility were obtained at values lower than 18% and 21%, respectively. The limit of quantification (LOQ) was established for most of the compounds at 1.0 μg/kg, except for testosterone in chicken and hydrocortisone-21-acetate and cortisone-21-acetate in pork at 2.0 μg/kg. Decision limit (CCα) and detection capability (CCβ) values ranged from 1.0-2.7 μg/kg and 1.9-5.5 μg/kg, respectively, in the three matrices. Finally, thirty one meat samples were analyzed and two hormones, progesterone and hydrocortisone, were detected in a beef and pork sample at 1.7 and 2.8 μg/kg respectively.
Sigma-Aldrich
Met-Arg-Phe-Ala acetate salt, ≥90% (HPLC)