Merck
  • Fatty Acids of CLA-Enriched Egg Yolks Can Induce Transcriptional Activation of Peroxisome Proliferator-Activated Receptors in MCF-7 Breast Cancer Cells.
Fatty Acids of CLA-Enriched Egg Yolks Can Induce Transcriptional Activation of Peroxisome Proliferator-Activated Receptors in MCF-7 Breast Cancer Cells.
PPAR research (2017-05-02)
Aneta A Koronowicz, Paula Banks, Adam Master, Dominik Domagała, Ewelina Piasna-Słupecka, Mariola Drozdowska, Elżbieta Sikora, Piotr Laidler
ABSTRACT
In our previous study, we showed that fatty acids from CLA-enriched egg yolks (EFA-CLA) reduced the proliferation of breast cancer cells; however, the molecular mechanisms of their action remain unknown. In the current study, we used MCF-7 breast cancer cell line to determine the effect of EFA-CLA, as potential ligands for peroxisome proliferator-activated receptors (PPARs), on identified in silico PPAR-responsive genes: BCAR3, TCF20, WT1, ZNF621, and THRB (transcript TRβ2). Our results showed that EFA-CLA act as PPAR ligands with agonistic activity for all PPAR isoforms, with the highest specificity towards PPARγ. In conclusion, we propose that EFA-CLA-mediated regulation of PPAR-responsive genes is most likely facilitated by cis9,trans11CLA isomer incorporated in egg yolk. Notably, EFA-CLA activated PPAR more efficiently than nonenriched FA as well as synthetic CLA isomers. We also propose that this regulation, at least in part, can be responsible for the observed reduction in the proliferation of MCF-7 cells treated with EFA-CLA.
Sigma-Aldrich
Anti-PPARG (AB1) antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-TCF20 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-PPARD antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-PPARA antibody produced in rabbit, affinity isolated antibody