Merck
Targeted Protein Degradation: from Chemical Biology to Drug Discovery.
Cell chemical biology (2017-06-27)
Philipp M Cromm, Craig M Crews
ABSTRACT
Traditional pharmaceutical drug discovery is almost exclusively focused on directly controlling protein activity to cure diseases. Modulators of protein activity, especially inhibitors, are developed and applied at high concentration to achieve maximal effects. Thereby, reduced bioavailability and off-target effects can hamper compound efficacy. Nucleic acid-based strategies that control protein function by affecting expression have emerged as an alternative. However, metabolic stability and broad bioavailability represent development hurdles that remain to be overcome for these approaches. More recently, utilizing the cell's own protein destruction machinery for selective degradation of essential drivers of human disorders has opened up a new and exciting area of drug discovery. Small-molecule-induced proteolysis of selected substrates offers the potential of reaching beyond the limitations of the current pharmaceutical paradigm to expand the druggable target space.
Sigma-Aldrich
(S,R,S)-AHPC-CO-PEG4-C2-acid, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-acetamido-O-PEG4-C1-acid, ≥95%
Sigma-Aldrich
Pomalidomide-piperazine, ≥ 95.0%
Sigma-Aldrich
(S,R,S)-AHPC-CO-PEG1-C2-acid, ≥95%
Sigma-Aldrich
Pomalidomide-C9-NH2 hydrochloride
Sigma-Aldrich
(S,R,S)-AHPC-PEG4-Alkyne
Sigma-Aldrich
Pomalidomide-PEG5-Alkyne, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG6-Azide, ≥95%
Sigma-Aldrich
Pomalidomide-PEG4-Azide, ≥95%
Sigma-Aldrich
Pomalidomide-C3-CO2H, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG1-Alkyne, ≥95%
Sigma-Aldrich
Pomalidomide-PEG5-NH2 hydrochloride
Sigma-Aldrich
2,7-Diazaspiro[3.5]nonane-7-acetic acid, 2-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-, ≥95%
Sigma-Aldrich
Thalidomide-O-amido-C8-NH2 trifluoroacetate, ≥95.0%
Sigma-Aldrich
Pomalidomide-PEG2-C2-azide, ≥95%
Sigma-Aldrich
VH 032 amide-alkyl C5-acid, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C6-PEG3-butyl amine hydrochloride, ≥95%
Sigma-Aldrich
4-Pentynoic acid, 5-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1-oxo-1H-isoindol-4-yl], ≥95.0%
Sigma-Aldrich
(S,R,S)-AHPC-PEG6-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG5-Alkyne
Sigma-Aldrich
Pomalidomide-PEG4-NH2 hydrochloride, ≥95%
Sigma-Aldrich
3-Pyridinecarboxylic acid, 6-[4-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-1-piperazinyl]-, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-piperazine-pyridine-alkyne-NH2 hydrochloride
Sigma-Aldrich
Propanoic acid, 3-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]amino]ethoxy]ethoxy]ethoxy]-, ≥95%
Sigma-Aldrich
Pomalidomide-C2-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-pentanoic-acid, ≥95%
Sigma-Aldrich
VH032-cyclopropane-F, ≥95%
Sigma-Aldrich
FBnG-C3-PEG5-C3-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C9-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-difluoroPEG1-C4-piperazine Hydrochloride, ≥95%