
Zopiclone R-Isomer

Synonym(s):
(R)-Zopiclone, (−)-(5R)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate
Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
Products
Brand
Formula Weight
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing