
Zinc acetate dihydrate

Synonym(s):
Zinc acetate dihydrate
Linear Formula:
(CH3COO)2Zn · 2 H2O
CAS Number:
Molecular Weight:
183.48 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing